Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-1H-pyrazole 98%

Supplier: AMBEED, INC
Danger

A463839-5G A463839-25G A463839-10G A463839-1G
103839-036EA 6 USD InStock
103839-036 76842-938 77116-318 77116-320
1-Methyl-1H-pyrazole 98%
1-Methyl-1H-pyrazole
Precaución: for Research Use Only


Formula: C₄H₆N₂
Boiling Pt: 127 °C (760 mm Hg)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00144943
CAS Number: 930-36-9
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty