Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Oxoisoindoline-5-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A149848-1G A149848-100MG A149848-250MG A149848-50MG A149848-1G
103804-090EA 507 USD InStock
103804-090 76823-806 76823-808 76823-810 77299-758
1-Oxoisoindoline-5-carbaldehyde 95%
1-Oxoisoindoline-5-carbaldehyde
Precaución: for Research Use Only


Formula: C₉H₇NO₂
Boiling Pt: 493.8±45.0 °C (760 mm Hg)
Flash Pt: 243.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13177813
CAS Number: 926307-99-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty