Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenyl-1,2,3,4-tetrahydroisoquinoline 97%

Supplier: AMBEED, INC
Warning

A297761-1G A297761-100G A297761-10G A297761-1G A297761-5G A297761-25G
103814-150EA 5 USD InStock
103814-150 103835-294 76787-656 76787-658 76837-978 76837-980 77305-718
1-Phenyl-1,2,3,4-tetrahydroisoquinoline 97%
1-Phenyl-1,2,3,4-tetrahydroisoquinoline
Precaución: for Research Use Only


Formula: C₁₅H₁₅N
Boiling Pt: 338.4 °C (760 mm Hg)
Melting Pt: 96…98 °C
Flash Pt: 166.9 °C
Storage Temperature: Ambient
MDL Number: MFCD02179241
CAS Number: 22990-19-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty