Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenyl-1,2-propanedione 98%

Supplier: AMBEED, INC
Warning

A171373-1G A171373-5G A171373-25G A171373-100G A171373-10G A171373-500G A171373-1G
103806-628EA 6 USD InStock
103806-628 103844-902 77082-054 77236-266 77236-268 77236-270 77301-126
1-Phenyl-1,2-propanedione 98%
1-Phenyl-1,2-propanedione
Precaución: for Research Use Only


Formula: C₆H₅C(O)C(O)CH₃
MW: 148.16 g/mol
Boiling Pt: 228
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00008755
CAS Number: 579-07-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty