Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenyl-1H-pyrazole 97%

Supplier: AMBEED, INC
Warning

A586768-5G A586768-1G A586768-25G A586768-5G A586768-100G A586768-10G
103828-898EA 5 USD InStock
103828-898 103840-044 76844-186 76844-188 76844-190 77242-462 77242-464
1-Phenyl-1H-pyrazole 97%
1-Phenyl-1H-pyrazole
Precaución: for Research Use Only


Formula: C₉H₈N₂
MW: 144.18
Boiling Pt: 251
Melting Pt: 11
Density: 1.091
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003112
CAS Number: 1126-00-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty