Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Propylimidazole 97%

Supplier: AMBEED, INC
Warning

A142246-100G A142246-500G A142246-5G A142246-25G
103808-628EA 283 USD InStock
103808-628 77137-366 77137-368 77137-370 77137-372 77316-346
1-Propylimidazole 97%
1-Propylimidazole
Precaución: for Research Use Only


Formula: C₆H₁₀N₂
Boiling Pt: 221.3 °C (760 mm Hg)
Flash Pt: 87.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00060494
CAS Number: 35203-44-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty