Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((4-(Carboxymethyl)phenoxy)methyl)benzoic acid 98%

Supplier: AMBEED, INC
Warning

A101268-1G A101268-5G A101268-100G A101268-250MG A101268-25G A101268-1G A101268-5G
103795-882EA 9 USD InStock
103795-882 103821-568 76869-138 76869-140 76869-142 77282-696 77297-036
2-((4-(Carboxymethyl)phenoxy)methyl)benzoic acid 98%
2-((4-(Carboxymethyl)phenoxy)methyl)benzoic acid
Precaución: for Research Use Only


Formula: C₁₆H₁₄O₅
Boiling Pt: 516.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD16619181
CAS Number: 55453-89-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty