Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((4-Cyanophenyl)amino)acetic acid 98%

Supplier: AMBEED, INC
Warning

A147588-5G A147588-100G A147588-500G A147588-25G A147588-5G
103841-910EA 5 USD InStock
103841-910 76719-596 76719-598 77317-294 77317-296
2-((4-Cyanophenyl)amino)acetic acid 98%
2-((4-Cyanophenyl)amino)acetic acid
Precaución: for Research Use Only


Formula: C₉H₈N₂O₂
Flash Pt: 224.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01464022
CAS Number: 42288-26-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty