Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1H-Imidazol-1-yl)acetonitrile 97%

Supplier: AMBEED, INC
Warning

A177742-1G A177742-5G A177742-100G A177742-10G A177742-1G A177742-25G A177742-5G A177742-500G
103807-266EA 6 USD InStock
103807-266 103845-598 76799-576 76799-578 77082-986 77082-988 77082-990 77879-173
2-(1H-Imidazol-1-yl)acetonitrile 97%
2-(1H-Imidazol-1-yl)acetonitrile
Precaución: for Research Use Only


Formula: C₅H₅N₃
Boiling Pt: 329.7 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD06421433
CAS Number: 98873-55-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty