Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indol-5-ylacetonitrile 95%

Supplier: AMBEED, INC
Warning

A198689-1G A198689-5G A198689-100MG A198689-250MG A198689-1G
103817-042EA 146 USD InStock
103817-042 103847-670 76975-162 76975-164 77085-694
1H-Indol-5-ylacetonitrile 95%
1H-Indol-5-ylacetonitrile
Precaución: for Research Use Only


Formula: C₁₀H₈N₂
MW: 156.184 g/mol
Boiling Pt: 374.1 °C (760 mm Hg)
Melting Pt: 58 °C
Flash Pt: 127.6 °C
Storage Temperature: Ambient
MDL Number: MFCD09263862
CAS Number: 23690-49-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty