Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Carbobenzoxy)glycylglycine 95%

Supplier: AMBEED, INC
Warning

A144650-1G A144650-5G A144650-100G A144650-10G A144650-1G A144650-25G A144650-5G A144650-500G
103803-430EA 6 USD InStock
103803-430 103841-508 76719-454 76719-456 77316-754 77316-756 77316-758 77879-020
(Carbobenzoxy)glycylglycine 95%
(Carbobenzoxy)glycylglycine
Precaución: for Research Use Only


Formula: C₁₂H₁₄N₂O₅
Boiling Pt: 587.2 °C (760 mm Hg)
Flash Pt: 309 °C
Storage Temperature: Ambient
MDL Number: MFCD00037783
CAS Number: 2566-19-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty