Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2,6-Dichlorophenoxy)acetic acid 97%

Supplier: AMBEED, INC
Warning

A102512-1G A102512-5G A102512-100MG A102512-250MG
103796-378EA 144 USD InStock
103796-378 103822-074 77245-192 77745-782
2-(2,6-Dichlorophenoxy)acetic acid 97%
2-(2,6-Dichlorophenoxy)acetic acid
Precaución: for Research Use Only


Formula: C₈H₆Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00272886
CAS Number: 575-90-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty