Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Acetylglycylglycine

Supplier: AMBEED, INC

A202743-1G A202743-5G A202743-25G A202743-100G A202743-10G A202743-5G
103817-412EA 30 USD InStock
103817-412 103848-056 103861-320 77112-210 77112-212 77302-822
N-Acetylglycylglycine
Proteins and Peptides
  • Boiling point: 599.6 °C (760 mm Hg)
  • Flash point: 316.4 °C
  • GHS symbol and signal word: GHS07 and warning
  • Melting point: 178 to 179 °C

Precaución: Sealed in dry, at 2 to 8 °C.


Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty