Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Aminoethyl)phenol 98%

Supplier: AMBEED, INC
Warning

A151404-1G A151404-5G A151404-1G A151404-5G A151404-100MG
103804-254EA 43 USD InStock
103804-254 103842-406 77317-968 77317-970 77748-047
2-(2-Aminoethyl)phenol 98%
2-(2-Aminoethyl)phenol
Precaución: for Research Use Only


Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 258 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00192585
CAS Number: 2039-66-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty