Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Aminophenyl)acetonitrile 97%

Supplier: AMBEED, INC
Warning

A118974-1G A118974-10G A118974-250MG A118974-1G A118974-5G
103799-142EA 29 USD InStock
103799-142 77246-018 77246-020 77311-806 77311-808
(2-Aminophenyl)acetonitrile 97%
(2-Aminophenyl)acetonitrile
Precaución: for Research Use Only


Formula: C₈H₈N₂
MW: 132.17 g/mol
Boiling Pt: 302.2 °C (760 mm Hg)
Melting Pt: 69…72 °C
Flash Pt: 136.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00091082
CAS Number: 2973-50-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty