Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Aminopropan-2-yl)-N-(4-fluorobenzyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidine-4-carboxamide 97%

Supplier: AMBEED, INC
Warning

A298987-1G A298987-5G A298987-10G A298987-250MG A298987-1G A298987-5G
103814-206EA 19 USD InStock
103814-206 103835-354 77032-448 77032-450 77305-758 77305-760
2-(2-Aminopropan-2-yl)-N-(4-fluorobenzyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidine-4-carboxamide 97%
2-(2-Aminopropan-2-yl)-N-(4-fluorobenzyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidine-4-carboxamide
Precaución: for Research Use Only


Formula: C₁₆H₁₉FN₄O₃
MW: 334.3500 g/mol
Boiling Pt: 567.4 °C (760 mm Hg)
Melting Pt: 105…107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10698741
CAS Number: 518048-03-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty