Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Aminopyridin-3-yl)acetic acid 95%

Supplier: AMBEED, INC
Warning

A147764-1G A147764-5G A147764-250MG A147764-100MG
103803-826EA 271 USD InStock
103803-826 103841-934 77137-638 77685-241
2-(2-Aminopyridin-3-yl)acetic acid 95%
2-(2-Aminopyridin-3-yl)acetic acid
Precaución: for Research Use Only


Formula: C₇H₈N₂O₂
Melting Pt: 219…221.5 °C
Storage Temperature: Ambient
MDL Number: MFCD11848318
CAS Number: 101860-97-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty