Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Bromoethoxy)propane 98%

Supplier: AMBEED, INC
Warning

A103121-1G A103121-5G A103121-100MG A103121-250MG A103121-1G A103121-5G
103796-484EA 57 USD InStock
103796-484 103822-188 76809-372 76809-374 77309-060 77309-062
2-(2-Bromoethoxy)propane 98%
2-(2-Bromoethoxy)propane
Precaución: for Research Use Only


Formula: C₅H₁₁BrO
MW: 167.044 g/mol
Boiling Pt: 148.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07700570
CAS Number: 54149-16-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty