Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Bromoethoxy)tetrahydropyran 95% stabilized

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with potassium carbonate

A194345-1G A194345-5G A194345-1G A194345-100G A194345-10G A194345-25G A194345-5G A194345-500G
103816-642EA 9 USD InStock
103816-642 103847-254 77085-172 77236-994 77236-996 77302-408 77302-410 77746-235
2-(2-Bromoethoxy)tetrahydropyran 95% stabilized
2-(2-Bromoethoxy)tetrahydropyran
Precaución: for Research Use Only


Formula: C₇H₁₃BrO₂
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD01321310
CAS Number: 17739-45-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty