Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Chloroethoxy)acetic acid 95%

Supplier: AMBEED, INC
Warning

A155072-1G A155072-5G A155072-100MG A155072-250MG
103804-712EA 85 USD InStock
103804-712 103842-908 76973-276 76973-278
(2-Chloroethoxy)acetic acid 95%
(2-Chloroethoxy)acetic acid
Precaución: for Research Use Only


Formula: C₄H₇ClO₃
MW: 138.55 g/mol
Boiling Pt: 289.8 °C (760 mm Hg)
Flash Pt: 129.038 °C
Storage Temperature: Ambient
MDL Number: MFCD17014832
CAS Number: 14869-41-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty