Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxyphenylboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A157679-1G A157679-5G A157679-100G A157679-250MG A157679-1G A157679-25G A157679-5G
103805-034EA 6 USD InStock
103805-034 103843-236 76719-982 76719-984 77319-072 77319-074 77319-076
2-Methoxyphenylboronic acid pinacol ester 97%
2-Methoxyphenylboronic acid pinacol ester


Formula: C₁₃H₁₉BO₃
Boiling Pt: 328.5±25.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD05863916
CAS Number: 190788-60-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty