Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3,4-Dichlorophenyl)ethanol 98%

Supplier: AMBEED, INC
Warning

A118485-1G A118485-25G A118485-5G A118485-1G
103799-066EA 14 USD InStock
103799-066 77271-362 77297-606 77311-738
2-(3,4-Dichlorophenyl)ethanol 98%
2-(3,4-Dichlorophenyl)ethanol
Precaución: for Research Use Only


Formula: C₈H₈Cl₂O
MW: 191.05 g/mol
Boiling Pt: 279.1 °C (760 mm Hg)
Flash Pt: 117.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00800673
CAS Number: 35364-79-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty