Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Amino-1H-pyrazol-1-yl)ethanol 97%

Supplier: AMBEED, INC
Warning

A101151-1G A101151-5G A101151-100MG A101151-1G A101151-5G
103795-824EA 33 USD InStock
103795-824 103821-514 76869-130 77297-000 77297-002
2-(3-Amino-1H-pyrazol-1-yl)ethanol 97%
2-(3-Amino-1H-pyrazol-1-yl)ethanol
Precaución: for Research Use Only


Formula: C₅H₉N₃O
MW: 127.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12149344
CAS Number: 84407-13-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty