Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Fluorophenyl)pyrrolidine 95%

Supplier: AMBEED, INC
Warning

A194895-1G A194895-5G A194895-100MG A194895-250MG
103816-704EA 57 USD InStock
103816-704 103847-318 76691-040 77877-520
2-(3-Fluorophenyl)pyrrolidine 95%
2-(3-Fluorophenyl)pyrrolidine
Precaución: for Research Use Only


Formula: C₁₀H₁₂FN
MW: 165.2074 g/mol
Boiling Pt: 232.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD02663475
CAS Number: 298690-72-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty