Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Carboxymethyl)phenylboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A328419-1G A328419-5G A328419-100MG A328419-250MG A328419-1G A328419-5G A328419-25G
103825-498EA 21 USD InStock
103825-498 103836-452 76724-086 76724-088 77306-376 77306-378 77879-692
4-(Carboxymethyl)phenylboronic acid pinacol ester 97%
4-(Carboxymethyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₁₉BO₄
Boiling Pt: 398.2 °C (760 mm Hg)
Melting Pt: 165…170 °C
Flash Pt: 194.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038755
CAS Number: 797755-07-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty