Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-(Methoxycarbonyl)phenyl)acetic acid 98%

Supplier: AMBEED, INC
Warning

A158946-1G A158946-5G A158946-100MG A158946-10G A158946-250MG A158946-1G A158946-25G
103805-200EA 19 USD InStock
103805-200 103843-412 76871-956 76871-958 76871-960 77319-274 77319-276
2-(4-(Methoxycarbonyl)phenyl)acetic acid 98%
2-(4-(Methoxycarbonyl)phenyl)acetic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₄
Boiling Pt: 349.6 °C (760 mm Hg)
Melting Pt: 112…115 °C
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD19440718
CAS Number: 22744-12-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty