Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetate 97%

Supplier: AMBEED, INC
Warning

A126322-1G A126322-100MG A126322-10G A126322-250MG A126322-25G A126322-1G A126322-5G A126322-100G
103799-322EA 42 USD InStock
103799-322 103800-792 77161-626 77161-628 77161-630 77161-632 77312-998 77313-000 77571-562
Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetate 97%
Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetate
Precaución: for Research Use Only


Formula: C₂₈H₅₂N₄O₈
MW: 572.74 g/mol
Boiling Pt: 632.127 °C (760 mm Hg)
Flash Pt: 336.099 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02259697
CAS Number: 137076-54-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty