Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 98%

Supplier: AMBEED, INC
Warning

A149055-1G A149055-5G A149055-100G A149055-10G A149055-1G A149055-25G
103803-996EA 5 USD InStock
103803-996 103842-116 76689-296 76689-298 77317-590 77317-592
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 98%
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol
Precaución: for Research Use Only


Formula: C₉H₇F₆NO
Boiling Pt: 292.9 °C (760 mm Hg)
Melting Pt: 147…152 °C
Flash Pt: 130.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039258
CAS Number: 722-92-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty