Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromophenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A118429-1G A118429-100G A118429-10G A118429-250MG A118429-25G A118429-1G A118429-5G
103799-054EA 6 USD InStock
103799-054 76822-270 76822-272 76822-274 76822-276 77311-726 77311-728
4-Bromophenylboronic acid pinacol ester 98%
4-Bromophenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₁₆BBrO₂
Boiling Pt: 324.1 °C (760 mm Hg)
Flash Pt: 149.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11110553
CAS Number: 68716-49-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty