Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorobenzylboronic acid pinacol ester 96%

Supplier: AMBEED, INC
Warning

A230449-1G A230449-5G A230449-250MG A230449-1G A230449-5G
103819-548EA 41 USD InStock
103819-548 103850-448 76874-236 77089-308 77089-310
4-Chlorobenzylboronic acid pinacol ester 96%
4-Chlorobenzylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₈BClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD10698531
CAS Number: 475250-49-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty