Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorophenylboronic acid pinacol ester 95%

Supplier: AMBEED, INC
Warning

A135150-1G A135150-100G A135150-10G A135150-25G A135150-1G A135150-5G
103802-098EA 9 USD InStock
103802-098 77285-472 77285-474 77285-476 77314-890 77314-892
4-Chlorophenylboronic acid pinacol ester 95%
4-Chlorophenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₁₆BClO₂
Boiling Pt: 308.678 °C (760 mm Hg)
Melting Pt: 50…55 °C
Flash Pt: 140.484 °C
Storage Temperature: Ambient
MDL Number: MFCD05663875
CAS Number: 195062-61-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty