Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isopropylphenylacetic acid 98%

Supplier: AMBEED, INC
Warning

A121239-1G A121239-10G A121239-250MG A121239-25G A121239-1G A121239-5G
103800-048EA 15 USD InStock
103800-048 76797-384 76797-386 76797-388 76797-390 77312-164 77312-166
4-Isopropylphenylacetic acid 98%
4-Isopropylphenylacetic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₂
Boiling Pt: 295 °C (760 mm Hg)
Melting Pt: 51…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00041035
CAS Number: 4476-28-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty