Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Methoxyphenyl)ethanol 98%

Supplier: AMBEED, INC
Warning

A213537-5G A213537-25G A213537-100G A213537-10G A213537-500G A213537-5G
103849-012EA 9 USD InStock
103849-012 77087-528 77288-024 77288-026 77288-028 77303-294
2-(4-Methoxyphenyl)ethanol 98%
2-(4-Methoxyphenyl)ethanol
Precaución: for Research Use Only


Formula: C₉H₁₂O₂
Boiling Pt: 334…336 °C (760 mm Hg)
Melting Pt: 26…28 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002900
CAS Number: 702-23-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty