Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(6-Bromopyridin-2-yl)acetic acid 97%

Supplier: AMBEED, INC
Warning

A243100-1G A243100-100MG A243100-250MG A243100-1G
103811-504EA 131 USD InStock
103811-504 77031-314 77031-316 77090-520
2-(6-Bromopyridin-2-yl)acetic acid 97%
2-(6-Bromopyridin-2-yl)acetic acid
Precaución: for Research Use Only


Formula: C₇H₆BrNO₂
Boiling Pt: 347.2±27.0 °C (760 mm Hg)
Flash Pt: 163.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10000075
CAS Number: 1093879-46-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty