Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(6-Bromopyridin-2-yl)benzaldehyde 95%

Supplier: AMBEED, INC
Warning

A105825-1G A105825-5G A105825-250MG A105825-1G A105825-5G
103796-952EA 26 USD InStock
103796-952 103822-696 76780-802 77309-560 77309-562
2-(6-Bromopyridin-2-yl)benzaldehyde 95%
2-(6-Bromopyridin-2-yl)benzaldehyde
Precaución: for Research Use Only


Formula: C₁₂H₈BrNO
MW: 262.1000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05864830
CAS Number: 914349-51-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty