Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(6-Hydroxybenzofuran-3-yl)acetic acid 95%

Supplier: AMBEED, INC
Warning

A262340-1G A262340-5G A262340-1G A262340-5G A262340-10G
103812-560EA 11 USD InStock
103812-560 103852-456 77092-074 77092-076 77238-532
2-(6-Hydroxybenzofuran-3-yl)acetic acid 95%
2-(6-Hydroxybenzofuran-3-yl)acetic acid
Precaución: for Research Use Only


Formula: C₁₀H₈O₄
Boiling Pt: 251.5 °C (760 mm Hg)
Melting Pt: 139…140 °C
Flash Pt: 105.9 °C
Storage Temperature: Ambient
MDL Number: MFCD05273635
CAS Number: 69716-04-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty