Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(7-(Dimethylamino)-2-oxo-2H-chromen-4-yl)acetic acid 98%

Supplier: AMBEED, INC
Warning

A101837-1G A101837-5G A101837-100MG A101837-250MG A101837-1G
103796-166EA 66 USD InStock
103796-166 103821-860 76687-054 76687-056 77308-752
2-(7-(Dimethylamino)-2-oxo-2H-chromen-4-yl)acetic acid 98%
2-(7-(Dimethylamino)-2-oxo-2H-chromen-4-yl)acetic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₃NO₄
Boiling Pt: 497.6 °C (760 mm Hg)
Melting Pt: 180…184 °C
Flash Pt: 254.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00857826
CAS Number: 80883-54-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty