Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(7-Amino-2-oxo-2H-chromen-4-yl)acetic acid 97%

Supplier: AMBEED, INC
Warning

A138357-1G A138357-100MG A138357-10G A138357-250MG A138357-1G A138357-25G A138357-5G
103802-604EA 55 USD InStock
103802-604 76782-792 76782-794 76782-796 77298-984 77298-986 77315-626
2-(7-Amino-2-oxo-2H-chromen-4-yl)acetic acid 97%
2-(7-Amino-2-oxo-2H-chromen-4-yl)acetic acid
Precaución: for Research Use Only


Formula: C₁₁H₉NO₄
Flash Pt: 269.052 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13185880
CAS Number: 85157-21-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty