Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyloxyacetyl chloride 95% (by GC)

Supplier: AMBEED, INC
Danger

A771958-10G A771958-25G A771958-5G A771958-1G
103840-938EA 22 USD InStock
103840-938 76697-832 76845-188 76845-190 77308-296
Benzyloxyacetyl chloride 95% (by GC)
Benzyloxyacetyl chloride
Precaución: for Research Use Only


Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Boiling Pt: 224 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010768
CAS Number: 19810-31-2
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty