Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Carboxymethyl)-5-fluorobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A272008-1G A272008-1G A272008-100MG A272008-250MG A272008-5G
103813-080EA 286 USD InStock
103813-080 77092-780 77140-996 77140-998 77141-000
2-(Carboxymethyl)-5-fluorobenzoic acid 97%
2-(Carboxymethyl)-5-fluorobenzoic acid
Precaución: for Research Use Only


Formula: C₉H₇FO₄
Storage Temperature: Refrigerator
MDL Number: MFCD19370105
CAS Number: 583880-95-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty