Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Chloromethyl)-3,4-dimethoxypyridine 1-oxide 95%

Supplier: AMBEED, INC
Danger

A1005671-1G A1005671-100MG A1005671-250MG
103795-514EA 298 USD InStock
103795-514 76646-402 76646-404
2-(Chloromethyl)-3,4-dimethoxypyridine 1-oxide 95%
2-(Chloromethyl)-3,4-dimethoxypyridine 1-oxide
Precaución: for Research Use Only


Formula: C₈H₁₀ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09840247
CAS Number: 953787-47-8
UN: 1759
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty