Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Chloromethyl)-5-methylpyridine hydrochloride 95%

Supplier: AMBEED, INC
Danger

A222323-1G A222323-5G A222323-5G A222323-100MG
103818-974EA 88 USD InStock
103818-974 103849-854 77303-624 77881-557
2-(Chloromethyl)-5-methylpyridine hydrochloride 95%
2-(Chloromethyl)-5-methylpyridine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₉Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD18253390
CAS Number: 71670-70-7
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty