Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Dimethylamino)acetaldehyde sulfite 97%

Supplier: AMBEED, INC
Warning

A120526-1G A120526-10G A120526-250MG A120526-25G A120526-1G
103799-950EA 15 USD InStock
103799-950 76983-220 76983-222 76983-224 77312-052
2-(Dimethylamino)acetaldehyde sulfite 97%
2-(Dimethylamino)acetaldehyde sulfite
Precaución: for Research Use Only


Formula: C₄H₁₁NO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD28139076
CAS Number: 1413945-87-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty