Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Furan-2-acetic acid 98%

Supplier: AMBEED, INC
Warning

A207637-1G A207637-5G A207637-100MG A207637-250MG A207637-5G
103817-812EA 79 USD InStock
103817-812 103848-494 76800-544 76800-546 77303-040
Furan-2-acetic acid 98%
Furan-2-acetic acid
Precaución: for Research Use Only


Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 233.8 °C (760 mm Hg)
Melting Pt: 68.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00798036
CAS Number: 2745-26-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty