Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Piperidinemethanol 98%

Supplier: AMBEED, INC
Warning

A179580-5G A179580-5G A179580-100G A179580-10G A179580-25G
103845-796EA 7 USD InStock
103845-796 77083-262 77273-982 77273-984 77301-620
2-Piperidinemethanol 98%
2-Piperidinemethanol
Precaución: for Research Use Only


Formula: C₆H₁₃NO
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00005987
CAS Number: 3433-37-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty