Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Methoxymethyl)benzene-1,4-diamine 95%

Supplier: AMBEED, INC
Warning

A144638-100MG A144638-250MG
103803-426EA 350 USD InStock
103803-426 103841-504 77190-562 77190-564
2-(Methoxymethyl)benzene-1,4-diamine 95%
2-(Methoxymethyl)benzene-1,4-diamine
Precaución: for Research Use Only


Formula: C₈H₁₂N₂O
MW: 152.19 g/mol
Boiling Pt: 296.0±25.0 °C (760 mm Hg)
Flash Pt: 150.2 °C
Storage Temperature: Ambient
MDL Number: MFCD20690892
CAS Number: 337906-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty