Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Methylamino)benzaldehyde 96%

Supplier: AMBEED, INC
Warning

A134352-1G A134352-100MG A134352-250MG A134352-1G
103802-006EA 71 USD InStock
103802-006 76983-896 76983-898 77314-748
2-(Methylamino)benzaldehyde 96%
2-(Methylamino)benzaldehyde
Precaución: for Research Use Only


Formula: C₈H₉NO
Boiling Pt: 260.7 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD19216814
CAS Number: 7755-70-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty