Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Naphthylacetonitrile 97%

Supplier: AMBEED, INC
Warning

A104510-5G A104510-100G A104510-10G A104510-25G A104510-5G A104510-500G
103822-442EA 5 USD InStock
103822-442 76994-740 76994-742 76994-744 77283-230 77670-510
1-Naphthylacetonitrile 97%
1-Naphthylacetonitrile
Precaución: for Research Use Only


Formula: C₁₂H₉N
Boiling Pt: 323.9 °C (760 mm Hg)
Melting Pt: 33…35 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004041
CAS Number: 132-75-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty