Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Phenylamino)propanoic acid 97%

Supplier: AMBEED, INC
Warning

A435768-1G A435768-100MG A435768-250MG A435768-5G A435768-1G
103827-604EA 427 USD InStock
103827-604 77168-724 77168-726 77168-728 77307-288
2-(Phenylamino)propanoic acid 97%
2-(Phenylamino)propanoic acid
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01940323
CAS Number: 15727-49-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty