Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Pyrimidyl)piperazine 98%

Supplier: AMBEED, INC
Warning

A372697-1G A372697-5G A372697-25G A372697-5G A372697-500G
103826-512EA 5 USD InStock
103826-512 103837-530 76840-790 76840-792 77626-722
1-(2-Pyrimidyl)piperazine 98%
1-(2-Pyrimidyl)piperazine
Precaución: for Research Use Only


Formula: C₈H₁₂N₄
Boiling Pt: 328 °C (760 mm Hg)
Melting Pt: 32…34 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00040742
CAS Number: 20980-22-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty